Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mycn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008709.3 | NP_032735.3 | 3 | 2175 | ||
GenBank mRNA | AK011628.1 | - | 1 | 518 | ||
AK144126.1 | - | 3 | 2175 | |||
AK146696.1 | - | 3 | 2176 | |||
AK146913.1 | - | 3 | 2174 | |||
BC005453.1 | - | 3 | 2055 | 3495446 | ||
BC049783.1 | - | 2 | 2101 | 6517457 |