Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004914.3 | NP_004905.2 | 6, 7 | 639 | ||
XM_005261859.4 | XP_005261916.1 | 7, 8 | 723 | |||
XM_006724381.3 | XP_006724444.1 | 7, 8 | 726 | |||
XM_006724382.3 | XP_006724445.1 | 6, 7 | 651 | |||
XM_011530544.2 | XP_011528846.1 | 7, 8 | 731 | |||
XM_011530545.2 | XP_011528847.1 | 7, 8 | 727 | |||
XM_011530546.2 | XP_011528848.1 | 7, 8 | 733 | |||
XM_011530547.2 | XP_011528849.1 | 5, 6 | 573 | |||
XM_017029104.1 | XP_016884593.1 | 6, 7 | 647 | |||
XM_017029105.1 | XP_016884594.1 | 5, 6 | 569 | |||
GenBank mRNA | AB023061.1 | - | 6, 7 | 639 | ||
AF133588.1 | - | 6, 7 | 609 | |||
AY336745.1 | - | 5, 6 | 533 | |||
BC104989.1 | - | 6, 7 | 628 | 8143992 | ||
BC104991.1 | - | 6, 7 | 628 | 8143994 | ||
CR456553.1 | - | 6, 7 | 617 |