Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290680.1 | NP_001277609.1 | 2 | 157 | ||
NM_001290681.1 | NP_001277610.1 | 2 | 153 | |||
NM_008741.4 | NP_032767.1 | 2 | 293 | |||
GenBank mRNA | AK011563.1 | - | 1 | 109 | ||
AK041338.1 | - | 2 | 211 | |||
AK049679.1 | - | 2 | 250 | |||
AK075612.1 | - | 2 | 250 | |||
AK088499.1 | - | 2 | 285 | |||
BC018224.1 | - | 2 | 225 | 4527900 | ||
BU058771.1 | - | 2 | 156 | |||
CD352422.1 | - | 2 | 153 | |||
DN176718.1 | - | 2 | 293 | |||
U17259.1 | - | 2 | 253 |