Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nthl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008743.2 | NP_032769.2 | 2 | 390 | ||
GenBank mRNA | AB006812.1 | - | 2 | 336 | ||
AK033701.1 | - | 2 | 1398 | |||
BC138852.1 | - | 2 | 353 | 8861874 | ||
BC138855.1 | - | 2 | 353 | 8861877 | ||
BC145443.1 | - | 2 | 262 | |||
CX565655.1 | - | 2 | 369 | |||
Y09688.1 | - | 2 | 362 |