Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008748.3 | NP_032774.1 | 5 | 1010 | ||
GenBank mRNA | BC052705.1 | - | 5 | 768 | 5697787 | |
X95518.1 | - | 4 | 653 |