Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163662.1 | NP_001157134.1 | 13 | 1475 | ||
NM_008749.2 | NP_032775.1 | 13 | 1463 | |||
GenBank mRNA | AK002630.1 | - | 13 | 1456 | ||
AK004886.1 | - | 13 | 1446 | |||
AK007976.1 | - | 13 | 1449 | |||
AK034944.1 | - | 169 | ||||
AK045087.1 | - | 13 | 1475 | |||
AK140710.1 | - | 13 | 1453 | |||
AK160981.1 | - | 13 | 1462 | |||
AK164088.1 | - | 13 | 1457 | |||
AK179888.1 | - | 2 | 156 | |||
AK215214.1 | - | 2 | 157 | |||
AK216538.1 | - | 2 | 157 | |||
BC072554.1 | - | 13 | 1453 | 5684979 | ||
M96823.1 | - | 12 | 1385 |