Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008750.5 | NP_032776.1 | 8 | 1704 | ||
GenBank mRNA | AK012352.1 | - | 8 | 1689 | ||
AK048109.1 | - | 8 | 1704 | |||
AK075628.1 | - | 2 | 591 | |||
AK149075.1 | - | 8 | 1688 | |||
BC004688.1 | - | 8 | 1569 | 3501314 | ||
BC047339.1 | - | 7 | 1141 | |||
BC058244.1 | - | 8 | 1704 | 6312910 |