Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004929.3 | NP_004920.1 | 4, 5 | 495 | ||
GenBank mRNA | AK054881.1 | - | 4, 5 | 493 | ||
AK295585.1 | - | 3, 4 | 214 | |||
AK312491.1 | - | 4, 5 | 493 | |||
AK315959.1 | - | 3, 4 | 201 | |||
BC006478.1 | - | 4, 5 | 483 | 2822391 | ||
BC020864.1 | - | 4, 5 | 493 | 4592497 | ||
HY128922.1 | - | 4, 5 | 496 | |||
X06661.1 | - | 4, 5 | 334 |