Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008786.2 | NP_032812.2 | 8 | 1200 | ||
GenBank mRNA | AK075632.1 | - | 7 | 687 | ||
AK179354.1 | - | 2 | 60 | |||
BC040750.1 | - | 8 | 746 | |||
BC049613.1 | - | 8 | 857 | |||
BC058966.1 | - | 8 | 889 | 5717963 | ||
D38023.1 | - | 8 | 864 | |||
M60320.1 | - | 8 | 864 | |||
S72473.1 | - | 2 | 145 |