Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282992.1 | NP_001269921.1 | 7, 8 | 1251 | ||
NM_008787.3 | NP_032813.3 | 7, 8 | 1251 | |||
GenBank mRNA | AB207233.1 | - | 7, 8 | 1125 | ||
AK145315.1 | - | 8, 9 | 1072 | |||
BC100380.1 | - | 7, 8 | 1046 | |||
BU518252.1 | - | 772 |