Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008795.2 | NP_032821.1 | 2 | 408 | ||
GenBank mRNA | AK004998.1 | - | 2 | 352 | ||
AK171569.1 | - | 2 | 189 | |||
BC119617.1 | - | 3 | 284 | 40061728 | ||
CJ070016.1 | - | 409 | ||||
X69026.1 | - | 2 | 425 |