Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008798.2 | NP_032824.1 | 5 | 1112 | ||
GenBank mRNA | AK039828.1 | - | 5 | 1092 | ||
BC119179.1 | - | 5 | 1081 | 8733928 | ||
BC120602.1 | - | 5 | 1081 | 40129525 | ||
X67914.1 | - | 5 | 1112 |