Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004950.4 | NP_004941.2 | 5 | 613 | ||
XM_011538008.1 | XP_011536310.1 | 4 | 429 | |||
GenBank mRNA | AK290642.1 | - | 5 | 570 | ||
BC030958.1 | - | 5 | 613 | 4604494 | ||
U59111.1 | - | 5 | 566 |