Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008818.2 | NP_032844.2 | 2 | 412 | ||
GenBank mRNA | AK146379.1 | - | 2 | 437 | ||
AK146393.1 | - | 2 | 436 | |||
AK146449.1 | - | 2 | 418 | |||
BC056204.1 | - | 3 | 420 | 30022528 | ||
DQ058642.1 | - | 2 | 396 | |||
M32484.1 | - | 2 | 396 |