Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161825.1 | NP_001155297.1 | 9 | 1343 | ||
NM_008822.2 | NP_032848.1 | 10 | 1421 | |||
GenBank mRNA | AK004687.1 | - | 10 | 1406 | ||
AK160195.1 | - | 10 | 1385 | |||
U69171.1 | - | 10 | 1413 |