Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162415.1 | NP_001155887.1 | 8, 9 | 751 | ||
NM_001162416.1 | NP_001155888.1 | 6, 7 | 544 | |||
NM_008825.4 | NP_032851.2 | 7, 8 | 645 | |||
GenBank mRNA | AK029659.1 | - | 8, 9 | 752 | ||
AK146744.1 | - | 7, 8 | 639 | |||
BC051014.1 | - | 6, 7 | 558 | 6493944 | ||
EU616667.1 | - | 6, 7 | 543 | |||
EU616668.1 | - | 6, 7 | 543 | |||
EU616669.1 | - | 6, 7 | 543 | |||
X98847.1 | - | 6, 7 | 563 |