Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008828.3 | NP_032854.2 | 8 | 946 | ||
GenBank mRNA | AK133877.1 | - | 8 | 868 | ||
AK145846.1 | - | 8 | 882 | |||
AK167441.1 | - | 8 | 840 | |||
AK167459.1 | - | 8 | 840 | |||
AK167710.1 | - | 8 | 839 | |||
AK177443.1 | - | 2 | 81 | |||
AK188712.1 | - | 1 | 12 | |||
AK207010.1 | - | 4 | 366 | |||
BC023819.1 | - | 8 | 852 | |||
BC032167.1 | - | 8 | 852 | |||
BC083355.1 | - | 8 | 838 | 6828087 | ||
BC108372.1 | - | 8 | 850 | 5060103 | ||
M15668.1 | - | 8 | 817 |