Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008829.2 | NP_032855.2 | 3, 4 | 2528 | ||
GenBank mRNA | AK036862.1 | - | 3, 4 | 1943 | ||
AK054117.1 | - | 3, 4 | 1209 | |||
AK142629.1 | - | 4, 5 | 850 | |||
BC145807.1 | - | 3, 4 | 1933 | 40131110 | ||
M68915.1 | - | 3, 4 | 1891 |