Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010571.3 | NP_034701.1 | 2 | 1094 | ||
GenBank mRNA | AF035948.1 | - | 2 | 900 | ||
AK030311.1 | - | 2 | 1096 | |||
AK040947.1 | - | 2 | 1079 | |||
BC098235.1 | - | 2 | 926 | 1546952 |