Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010575.2 | NP_034705.2 | 12 | 1322 | ||
GenBank mRNA | AF166384.1 | - | 12 | 1200 | ||
AF170316.1 | - | 12 | 1183 | |||
AK048700.1 | - | 7 | 830 | |||
AK140749.1 | - | 7 | 974 | |||
AK142289.1 | - | 12 | 1184 | |||
AK154619.1 | - | 12 | 1323 | |||
AK156872.1 | - | 11 | 1504 | |||
AK157012.1 | - | 11 | 1506 | |||
AK158443.1 | - | 12 | 1169 | |||
BC120493.1 | - | 12 | 1172 | 40129416 | ||
BC137570.1 | - | 12 | 1172 | 8860589 |