Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010575.2 | NP_034705.2 | 7 | 950 | ||
GenBank mRNA | AF166384.1 | - | 7 | 828 | ||
AF170316.1 | - | 7 | 811 | |||
AK048700.1 | - | 3 | 305 | |||
AK140749.1 | - | 3 | 283 | |||
AK142289.1 | - | 7 | 812 | |||
AK154619.1 | - | 7 | 951 | |||
AK156872.1 | - | 7 | 813 | |||
AK157012.1 | - | 7 | 812 | |||
AK158443.1 | - | 7 | 797 | |||
BC120493.1 | - | 7 | 800 | 40129416 | ||
BC137570.1 | - | 7 | 800 | 8860589 |