Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010577.3 | NP_034707.3 | 13 | 1505 | ||
GenBank mRNA | AK142241.1 | - | 13 | 1348 | ||
AK144921.1 | - | 13 | 1291 | |||
AK148138.1 | - | 13 | 1346 | |||
AK152594.1 | - | 13 | 1303 | |||
AK218419.1 | - | 2 | 252 | |||
BC050943.1 | - | 13 | 1521 | 6821577 | ||
BC053431.1 | - | 14 | 1566 | |||
BC059829.1 | - | 13 | 1521 | 6821577 | ||
X79003.1 | - | 13 | 1462 |