Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289450.1 | NP_001276379.1 | 15 | 1136 | ||
NM_010597.4 | NP_034727.3 | 14 | 1552 | |||
GenBank mRNA | AF033003.1 | - | 14 | 1907 | ||
AK004666.1 | - | 15 | 1120 | |||
AK015412.1 | - | 16 | 1392 | |||
AK138467.1 | - | 14 | 1541 | |||
AK153200.1 | - | 15 | 1138 | |||
BC014701.1 | - | 14 | 1277 | 4482268 | ||
U65591.1 | - | 14 | 1148 | |||
X97281.1 | - | 14 | 1536 |