Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267593.1 | NP_001254522.1 | 2 | 2150 | ||
NM_010603.6 | NP_034733.4 | 3 | 2322 | |||
GenBank mRNA | AK017299.1 | - | 3 | 2323 | ||
AK162389.1 | - | 1 | 1222 |