Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010605.4 | NP_034735.3 | 4 | 1399 | ||
GenBank mRNA | AB019560.1 | - | 2 | 940 | ||
AK052891.1 | - | 3 | 461 | |||
AK137600.1 | - | 4 | 1513 | |||
AK164058.1 | - | 4 | 1400 | |||
BC104330.1 | - | 2 | 958 | 40045462 | ||
BC104331.1 | - | 2 | 958 | 40045465 | ||
BC114976.1 | - | 2 | 955 | 40045466 | ||
U33631.1 | - | 2 | 960 |