Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010617.2 | NP_034747.2 | 8 | 711 | ||
GenBank mRNA | AB001429.1 | - | 3 | 323 | ||
AB037923.2 | - | 8 | 712 | |||
AK077053.1 | - | 8 | 734 |