Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190978.1 | NP_001177907.1 | 12 | 2498 | ||
NM_010623.4 | NP_034753.1 | 15 | 3330 | |||
GenBank mRNA | AB008867.1 | - | 15 | 3328 | ||
AK122506.1 | - | 5 | 3731 |