Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001715.2 | NP_001706.2 | 5 | 882 | ||
XM_011543824.1 | XP_011542126.1 | 5 | 903 | |||
XM_011543825.2 | XP_011542127.1 | 5 | 648 | |||
XM_011543827.1 | XP_011542129.1 | 4 | 779 | |||
XM_011543828.2 | XP_011542130.1 | 5 | 903 | |||
XM_011543829.2 | XP_011542131.1 | 5 | 903 | |||
XM_017013730.1 | XP_016869219.1 | 4 | 779 | |||
GenBank mRNA | AK313751.1 | - | 5 | 471 | ||
BC007371.2 | - | 5 | 445 | 3634568 | ||
BC018703.1 | - | 5 | 746 | |||
BC032413.1 | - | 5 | 491 | 5216030 | ||
S76617.1 | - | 5 | 889 | |||
Z33998.1 | - | 5 | 523 |