Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhmk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010633.3 | NP_034763.3 | 4, 5 | 998 | ||
GenBank mRNA | AK013347.1 | - | 4, 5 | 1003 | ||
AK030152.1 | - | 815 | ||||
BC058732.1 | - | 4, 5 | 998 | 6414877 | ||
X82320.1 | - | 3, 4 | 484 | |||
Y10725.2 | - | 4, 5 | 855 |