Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mafb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010658.3 | NP_034788.1 | 1 | 472 | ||
GenBank mRNA | AK132425.1 | - | 1 | 473 | ||
AK143209.1 | - | 1 | 474 | |||
BC016434.1 | - | 1 | 175 | 4485468 | ||
BC038256.1 | - | 1 | 420 | 5097650 | ||
L36435.1 | - | 1 | 252 |