Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mafb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010658.3 | NP_034788.1 | 1 | 2799 | ||
GenBank mRNA | AK143209.1 | - | 1 | 2801 | ||
BC038256.1 | - | 1 | 2751 | 5097650 |