Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010664.2 | NP_034794.2 | 6 | 1143 | ||
GenBank mRNA | AK145413.1 | - | 6 | 1133 | ||
AK167072.1 | - | 6 | 1132 | |||
AK167265.1 | - | 6 | 1127 | |||
AK167432.1 | - | 6 | 1130 | |||
AK167469.1 | - | 6 | 1133 | |||
AK167676.1 | - | 6 | 1132 | |||
BC020474.1 | - | 6 | 1133 | |||
BC089022.1 | - | 6 | 1105 | 30701486 | ||
M11686.1 | - | 6 | 1134 | |||
M36376.1 | - | 6 | 1129 |