Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap12-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010670.1 | NP_034800.1 | 1 | 508 | ||
GenBank mRNA | BC107319.1 | - | 1 | 495 | 40053395 | |
BC107320.1 | - | 1 | 495 | 40053396 |