Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278645.1 | NP_001265574.1 | 2 | 262 | ||
NM_001278646.1 | NP_001265575.1 | 2 | 295 | |||
NM_001311168.1 | NP_001298097.1 | 2 | 264 | |||
NM_005005.2 | NP_004996.1 | 2 | 297 | |||
GenBank mRNA | AF044956.1 | - | 2 | 291 | ||
AF067168.1 | - | 2 | 291 | |||
AF261090.1 | - | 2 | 296 | |||
AI907018.1 | - | 2 | 314 | |||
AK026538.1 | - | 2 | 280 | |||
AK302286.1 | - | 2 | 259 | |||
AK313432.1 | - | 2 | 259 | |||
BC007672.2 | - | 2 | 264 | 3160562 | ||
BQ685604.1 | - | 2 | 247 | |||
CN410734.1 | - | 2 | 205 | |||
DB504123.1 | - | 2 | 222 |