Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010696.3 | NP_034826.2 | 4 | 386 | ||
GenBank mRNA | AK036727.1 | - | 4 | 444 | ||
AK037970.1 | - | 4 | 420 | |||
AK088552.1 | - | 4 | 508 | |||
AK153374.1 | - | 4 | 391 | |||
AK168134.1 | - | 4 | 402 | |||
AK170491.1 | - | 4 | 663 | |||
BC006948.1 | - | 4 | 386 | 3603181 | ||
U20159.1 | - | 4 | 425 |