Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145953.1 | NP_001139425.1 | 6 | 1237 | ||
NM_010705.3 | NP_034835.1 | 6 | 1210 | |||
GenBank mRNA | AK089249.1 | - | 6 | 1154 | ||
BC132328.1 | - | 6 | 1163 | 40130605 | ||
BC138790.1 | - | 6 | 1163 | 8861812 | ||
BC145419.1 | - | 6 | 1163 | 9053977 | ||
BY440035.1 | - | 1 | 237 |