Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNU13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003796.1 | NP_001003796.1 | 3 | 553 | ||
NM_005008.3 | NP_004999.1 | 4 | 785 | |||
GenBank mRNA | AF091076.1 | - | 2 | 455 | ||
AF155235.1 | - | 3 | 516 | |||
BC005358.1 | - | 3 | 491 | 3681386 | ||
BC019282.2 | - | 2 | 441 | |||
BC095439.1 | - | 3 | 522 | 30719732 | ||
BG164621.1 | - | 3 | 633 | |||
CF552526.1 | - | 3 | 553 | |||
CR456531.1 | - | 3 | 469 | |||
D50420.1 | - | 3 | 534 |
Gene Symbol: ANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309356.1 | - | 472 |