Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136070.1 | NP_001129542.1 | 13 | 1588 | ||
NM_010709.3 | NP_034839.2 | 13 | 1588 | |||
GenBank mRNA | AK030312.1 | - | 13 | 1599 | ||
AK034115.1 | - | 13 | 1560 | |||
AK036154.1 | - | 13 | 1584 | |||
AK046539.1 | - | 13 | 1600 | |||
AK047005.1 | - | 12 | 1433 | |||
AK082748.1 | - | 13 | 1603 | |||
AK083638.1 | - | 13 | 1548 | |||
AK139052.1 | - | 13 | 1588 | |||
AK161269.1 | - | 13 | 1571 | |||
AK166085.1 | - | 13 | 1601 | |||
AK167980.1 | - | 13 | 1584 | |||
AK168120.1 | - | 13 | 1554 | |||
AK170252.1 | - | 13 | 1615 | |||
AK170448.1 | - | 13 | 1600 | |||
AK171115.1 | - | 13 | 1566 | |||
BC023709.1 | - | 13 | 1541 | |||
BC024501.1 | - | 8 | 902 | |||
BC025036.1 | - | 13 | 1544 | 4935223 | ||
U58337.1 | - | 13 | 1473 |