Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010712.2 | NP_034842.2 | 3, 4 | 602 | ||
GenBank mRNA | AB221565.1 | - | 2, 3 | 383 | ||
AF135415.1 | - | 2, 3 | 387 | |||
BC049834.1 | - | 3, 4 | 602 | 6493203 | ||
U89343.1 | - | 2, 3 | 365 |