Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010712.2 | NP_034842.2 | 6 | 1456 | ||
GenBank mRNA | AF135415.1 | - | 5 | 1241 | ||
BC049834.1 | - | 6 | 1456 | 6493203 | ||
S71659.1 | - | 3 | 826 |