Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291245.1 | NP_001278174.1 | 20 | 3305 | ||
NM_001291246.1 | NP_001278175.1 | 20 | 3302 | |||
NM_001291247.1 | NP_001278176.1 | 20 | 3306 | |||
NM_010716.3 | NP_034846.2 | 20 | 3309 | |||
GenBank mRNA | AK031540.1 | - | 20 | 3246 | ||
AK143531.1 | - | 20 | 3175 | |||
AK150644.1 | - | 20 | 3214 | |||
AK156018.1 | - | 20 | 3224 | |||
AK156030.1 | - | 9 | 1291 | |||
AK156825.1 | - | 9 | 1291 | |||
AK169648.1 | - | 1 | 43 | |||
AK181064.1 | - | 1 | 128 | |||
BC049240.1 | - | 20 | 3212 | 4504384 | ||
BC083500.1 | - | 20 | 3208 | 3673103 | ||
U66058.1 | - | 20 | 3243 |