Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010725.2 | NP_034855.2 | 4, 5 | 680 | ||
GenBank mRNA | AB221651.1 | - | 4, 5 | 680 | ||
AF078166.1 | - | 4, 5 | 680 | |||
BC119169.1 | - | 4, 5 | 759 | 8733918 | ||
BC125469.1 | - | 4, 5 | 759 | 40129984 |