Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010739.2 | NP_034869.1 | 13 | 2092 | ||
GenBank mRNA | AK132274.1 | - | 12 | 1938 | ||
BC024321.1 | - | 12 | 2058 | 4219376 | ||
J04634.1 | - | 13 | 2092 |