Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010744.3 | NP_034874.2 | 4 | 555 | ||
GenBank mRNA | AK133513.1 | - | 4 | 555 | ||
AK160525.1 | - | 3 | 458 | |||
AK166092.1 | - | 3 | 457 | |||
BC006715.1 | - | 3 | 449 | 3154568 | ||
BC016568.1 | - | 3 | 695 | |||
BC138498.1 | - | 4 | 603 | 8861518 | ||
BC138499.1 | - | 4 | 602 | 8861519 | ||
BC145298.1 | - | 4 | 568 | |||
U41805.1 | - | 4 | 531 |