Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039960.2 | NP_001035049.1 | 18, 19 | 2619 | ||
NM_001258401.2 | NP_001245330.1 | 18, 19 | 2565 | |||
XM_011539010.1 | XP_011537312.1 | 18, 19 | 2620 | |||
XM_011539011.1 | XP_011537313.1 | 18, 19 | 2565 | |||
XM_011539014.2 | XP_011537316.1 | 8, 9 | 1218 | |||
XM_017020240.1 | XP_016875729.1 | 18, 19 | 2647 | |||
XM_017020241.1 | XP_016875730.1 | 18, 19 | 2979 | |||
XM_017020242.1 | XP_016875731.1 | 18, 19 | 2634 | |||
GenBank mRNA | AB018282.1 | - | 20, 21 | 2699 | ||
AF069512.1 | - | 18, 19 | 2456 | |||
DQ063579.2 | - | 18, 19 | 2608 | |||
DQ975204.1 | - | 18, 19 | 2435 | |||
DQ996398.1 | - | 18, 19 | 2669 | |||
DQ996537.1 | - | 18, 19 | 2435 |