Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010748.2 | NP_034878.2 | 5 | 620 | ||
GenBank mRNA | AK089635.1 | - | 3 | 381 | ||
AK165585.1 | - | 5 | 635 | |||
L77884.1 | - | 5 | 620 | |||
U70015.1 | - | 5 | 620 |