Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010749.6 | NP_034879.2 | 2 | 289 | ||
GenBank mRNA | AK004656.1 | - | 2 | 301 | ||
AK133943.1 | - | 2 | 295 | |||
AK145913.1 | - | 2 | 301 | |||
AK149177.1 | - | 2 | 319 | |||
AK151607.1 | - | 2 | 350 | |||
AK153140.1 | - | 2 | 301 | |||
AK153302.1 | - | 2 | 370 | |||
AK153474.1 | - | 2 | 310 | |||
BC027210.1 | - | 2 | 244 | |||
BC046956.1 | - | 2 | 289 | |||
BC080811.1 | - | 2 | 289 | 6438831 | ||
M63286.1 | - | 2 | 197 | |||
X56831.1 | - | 2 | 229 | |||
X64068.1 | - | 2 | 292 |