Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010749.6 | NP_034879.2 | 2 | 321 | ||
GenBank mRNA | AK004656.1 | - | 2 | 333 | ||
AK133943.1 | - | 2 | 327 | |||
AK145913.1 | - | 2 | 333 | |||
AK149177.1 | - | 2 | 351 | |||
AK151607.1 | - | 2 | 382 | |||
AK153140.1 | - | 2 | 333 | |||
AK153302.1 | - | 2 | 402 | |||
AK153474.1 | - | 2 | 342 | |||
BC027210.1 | - | 2 | 276 | |||
BC046956.1 | - | 2 | 321 | |||
BC080811.1 | - | 2 | 321 | 6438831 | ||
M63286.1 | - | 2 | 229 | |||
X56831.1 | - | 2 | 261 | |||
X64068.1 | - | 2 | 324 |