Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252481.1 | NP_001239410.1 | 2 | 619 | ||
NM_010754.5 | NP_034884.2 | 2 | 508 | |||
GenBank mRNA | AF005743.1 | - | 1 | 236 | ||
AK007817.1 | - | 2 | 285 | |||
AY334552.1 | - | 1 | 177 | |||
BC021342.1 | - | 2 | 307 | 5066237 | ||
BC089184.1 | - | 2 | 619 | 5322611 | ||
BU058005.1 | - | 2 | 539 | |||
U60530.1 | - | 1 | 236 |