Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145835.1 | - | 3 | 1201 | ||
AK146043.1 | - | 3 | 1170 | |||
BC022952.1 | - | 3 | 1092 | 5132212 |